Palmitoleic acid, 99%
Palmitoleic acid, 99%
Palmitoleic acid, 99%
Thermo Scientific Chemicals

Palmitoleic acid, 99%

CAS: 373-49-9 | C16H30O2 | 254.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 376910010
Price (USD)
171.65
Special offer
Online exclusive
Ends: 28-Dec-2026
202.00
Save 30.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
171.65
Special offer
Online exclusive
Ends: 28-Dec-2026
202.00
Save 30.35
Each
Chemical Identifiers
CAS373-49-9
IUPAC Name(9Z)-hexadec-9-enoic acid
Molecular FormulaC16H30O2
InChI KeySECPZKHBENQXJG-FPLPWBNLSA-N
SMILESCCCCCC\C=C/CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4570 to 1.4590 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Clear liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only