3-Formylsalicylic acid hydrate, 97%
3-Formylsalicylic acid hydrate, 97%
3-Formylsalicylic acid hydrate, 97%
3-Formylsalicylic acid hydrate, 97%
3-Formylsalicylic acid hydrate, 97%
Thermo Scientific Chemicals

3-Formylsalicylic acid hydrate, 97%

CAS: 1266615-78-4 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 377070010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1266615-78-4
IUPAC Name3-formyl-2-hydroxybenzoic acid
Molecular FormulaC8H6O4
InChI KeyYOEUNKPREOJHBW-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Loss on drying=<11 % (1 g, 105°C, 3 hrs)
Appearance (Color)Yellow to beige to light brown
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only