ART™ Non-Filtered Standard Hinged Rack Pipette Tips
Actual product may vary
ART™ Non-Filtered Standard Hinged Rack Pipette Tips
Access tips one-handed with this versatile hinged rack, which is designed for the way you work.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleFor Use With (Equipment)Fit for Purpose
3772-05-HR300 μLLow RetentionUniversal FitCertified Quality
3101-05-HR1000 μLLow RetentionUniversal FitSterile
3102-05-HR1000 μLLow RetentionUniversal FitCertified Quality
3101-HR1000 μLStandardUniversal FitSterile
3772-HR300 μLStandardUniversal FitCertified Quality
3551-05-HR200 μLLow RetentionUniversal FitSterile
3552-05-HR200 μLLow RetentionUniversal FitCertified Quality
3552-HR200 μLStandardUniversal FitCertified Quality
3521-05-HR20 μLLow RetentionEppendorfSterile
3522-05-HR20 μLLow RetentionEppendorfCertified Quality
3522-HR20 μLStandardEppendorfCertified Quality
3511-HR10 μLExtended LengthUniversal FitSterile
3502-05-HR10 μLLow RetentionUniversal FitCertified Quality
Catalog number 3772-05-HR
HSN 84799090
Price (INR)
9,007.00
PK
Volume (Metric):
300 μL
Tip Style:
Low Retention
For Use With (Equipment):
Universal Fit
Fit for Purpose:
Certified Quality
Price (INR)
9,007.00
PK
Access tips with one hand using Thermo ScientificTM ARTTM Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.
Features:
  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Sterile tips are DNase, RNase, DNA, ATP, and Endotoxin Free
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Specifications
DescriptionART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric)300 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)2.3 in.
Length (Metric)5.84 cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatHinged Rack
Tip StyleLow Retention
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePK