Benzyl (triphenylphosphoranylidene)acetate, 97%
Benzyl (triphenylphosphoranylidene)acetate, 97%
Benzyl (triphenylphosphoranylidene)acetate, 97%
Thermo Scientific Chemicals

Benzyl (triphenylphosphoranylidene)acetate, 97%

CAS: 15097-38-8 | C27H23O2P | 410.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 377220050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS15097-38-8
IUPAC Namebenzyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular FormulaC27H23O2P
InChI KeyINKMLGJBBDRIQR-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Melting point120°C to 127°C
Appearance (Color)White to off-white
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only