4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™
4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™
4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™
Thermo Scientific Chemicals

4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™

CAS: 873-77-8 | C6H4BrClMg | 215.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 377258000
Price (TWD)
-
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS873-77-8
IUPAC Namebromo(4-chlorophenyl)magnesium
Molecular FormulaC6H4BrClMg
InChI KeyCDEMHJCJMMOFMB-UHFFFAOYSA-M
SMILESClC1=CC=C([Mg]Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green to brown
Titration Active base21.0 to 25.0 %
Appearance (Form)Slightly hazy solution
Titration Active base0.95 to 1.15 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only