Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Thermo Scientific™

Nunc™ Biobanking and Cell Culture Cryogenic Tubes

Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeCapacity (Metric)Thread StyleStyleUnit Size
377267PKRound (Self-standing)1.8 mLInternalWriting Surface, A-R BasePack of 450
374080Conical (Self-standing)0.5 mLInternalNo Writing Surface, A-R BaseCase of 960
374081Conical (Self-standing)1 mLInternalWriting SurfaceCase of 960
375353Conical (Self-standing)1 mLExternalWriting Surface, A-R BaseCase of 2000
375353PK
also known as 375353
Conical (Self-standing)1 mLExternalWriting Surface, A-R BasePack of 500
347597Conical (Self-standing)1 mLExternalNo Writing Surface, A-R BaseCase of 2000
347597PK
also known as 347597
Conical (Self-standing)1 mLExternalNo Writing Surface, A-R BasePack of 500
377224Conical (Self-standing)1 mLInternalWriting Surface, A-R BaseCase of 2000
377224PKConical (Self-standing)1 mLInternalWriting Surface, A-R BasePack of 500
366656Conical (Self-standing)1 mLInternalWriting SurfaceCase of 2000
366656PK
also known as 366656
Conical (Self-standing)1 mLInternalWriting SurfacePack of 500
375418Round (Self-standing)1.8 mLExternalWriting Surface, A-R BaseCase of 1800
375418PK
also known as 375418
Round (Self-standing)1.8 mLExternalWriting Surface, A-R BasePack of 450
347627Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R BaseCase of 1800
347627PK
also known as 347627
Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R BasePack of 450
377267Round (Self-standing)1.8 mLInternalWriting Surface, A-R BaseCase of 1800
368632Round (Self-standing)1.8 mLInternalWriting SurfaceCase of 1800
368632PKRound (Self-standing)1.8 mLInternalWriting SurfacePack of 450
363401Round1.8 mLInternalWriting SurfaceCase of 2000
363401PK
also known as 363401
Round1.8 mLInternalWriting SurfacePack of 500
351934Round1.8 mLInternalWriting Surface, No CapCase of 2500
379189Round (Self-standing)3.6 mLInternalWriting Surface, A-R BaseCase of 1600
379189PK
also known as 379189
Round (Self-standing)3.6 mLInternalWriting Surface, A-R BasePack of 400
366524Round3.6 mLInternalWriting SurfaceCase of 1600
366524PK
also known as 366524
Round3.6 mLInternalWriting SurfacePack of 400
367997Round3.6 mLInternalWriting Surface, No CapCase of 1500
337516Round (Self-standing)4.5 mLExternalWriting Surface, A-R BaseCase of 1200
337516PK
also known as 337516
Round (Self-standing)4.5 mLExternalWriting Surface, A-R BasePack of 300
347643Round (Self-standing)4.5 mLExternalNo Writing Surface, A-R BaseCase of 1200
347643PK
also known as 347643
Round (Self-standing)4.5 mLExternalNo Writing Surface, A-R BasePack of 300
379146Conical (Self-standing)4.5 mLInternalWriting Surface, A-R BaseCase of 1200
379146PK
also known as 379146
Conical (Self-standing)4.5 mLInternalWriting Surface, A-R BasePack of 300
363452Round4.5 mLInternalWriting SurfaceCase of 1200
363452PK
also known as 363452
Round4.5 mLInternalWriting SurfacePack of 300
Catalog number 377267PK
Price (USD)
413.86
Online Exclusive
486.90
Save 73.04 (15%)
Pack of 450
Bottom Shape:
Round (Self-standing)
Capacity (Metric):
1.8 mL
Thread Style:
Internal
Style:
Writing Surface, A-R Base
Unit Size:
Pack of 450
Price (USD)
413.86
Online Exclusive
486.90
Save 73.04 (15%)
Pack of 450
Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.
Features:
  • Non-cytotoxic, non-mutagenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Specifications
Anti-RotationYes
Capacity (English)0.0608 oz.
Closure MaterialPolypropylene
Closure TypeScrew Cap
Dnase FreeYes
Diameter (English) Outer0.492 in.
Diameter (Metric)12.5 mm
Length (English)1.929 in.
Length (Metric)49 mm
Packaging450 Tubes
Packaging Quantity450 Pk.
PrintedYes
Product LineNunc
RNase FreeYes
StyleWriting Surface, A-R Base
Vapor LN2 (to -150°C)Yes
Bottom ShapeRound (Self-standing)
Capacity (Metric)1.8 mL
MaterialPolypropylene
Product TypeCryogenic Storage Tube
SterilitySterile
Thread StyleInternal
Unit SizePack of 450

Frequently asked questions (FAQs)

Will the heat required to properly seal Nunc CryoFlex Tube Wrap adversely affect my cells?

Nunc Biobanking and Cell Culture Cryogenic Tubes are normally placed in a 4 degrees C ice bath during cell inoculation in the cryogenic tube. The heat required to shrink the polyethylene Nunc CryoFlex Tube Wrap will not raise the internal temperature enough to damage the cells. At this stage, cryoprotective agents are added.

What protocol should I follow for thawing cells, if my Nunc Biobanking and Cell Culture Cryogenic Tubes were previously immersed in liquid nitrogen without Nunc CryoFlex Tube Wrap?

We recommend the following protocol for thawing cells immersed in liquid nitrogen without Nunc CryoFlex Tube Wrap:
- Wear appropriate protective clothing including gloves, face guard, etc., during all transfer processes.
- The Nunc Biobanking and Cell Culture Cryogenic Tubes should be stored at least 24 hr in the gas phase of the liquid nitrogen freezer. This should allow trapped liquid nitrogen to evaporate and escape slowly, thereby reducing the possible pressure in the cryogenic tube.
- After at least 24 hr storage in the gas phase, the cryogenic tube may be put into a water bath and covered with a towel.
- After thawing, the contents of the cryogenic tube can be transferred.

Can I immerse Nunc Biobanking and Cell Culture Cryogenic Tubes in liquid nitrogen (liquid phase) for storage?

Of the Nunc Biobanking and Cell Culture Cryogenic Tubes, we recommend the internally threaded design, incorporating a silicon gasket, for liquid nitrogen immersion. At ultra low temperatures, any sealing material becomes inflexible. A theoretical risk of leakage is present when any mechanical closure is used. To facilitate leakproof storage, mechanically closed containers must be hermetically sealed in a protective envelope.

The Nunc cryo products includes Nunc CryoFlex Tube Wrap, a heat sealable polyethylene tubing designed for cryogenic tube encasement. The use of cryogenic tubes for freezing in liquid nitrogen (liquid phase) storage without CryoFlex tubing poses a safety hazard.
WARNING: Do not use Nunc Biobanking and Cell Culture Cryogenic Tubes in liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex Tube Wrap (Cat. No. 343958). Improper use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up, resulting in possible explosion or biohazard release. Use appropriate safety procedures as outlined in the Nalge Nunc International Cryopreservation Manual when handling and disposing of the vials.

What is the difference between your Nunc Biobanking and Cell Culture Cryogenic Tubes with internal threads and external threads?

Internally threaded Nunc Biobanking and Cell Culture Tubes are designed for vapor phase liquid nitrogen freezing. The cap incorporates a silicon gasket to enhance sealing properties.
Externally threaded Nunc Biobanking and Cell Culture Tubes are designed for mechanical freezing. The external screw cap reduces risk of contamination due to handling.

Can I autoclave Nunc Biobanking and Cell Culture Cryogenic Tubes?

We do not recommend autoclaving Nunc Biobanking and Cell Culture Tubes, even though they are made of polypropylene with polyethylene caps and stoppers.