Triethylaluminium, 1.3M solution in Heptane, Thermo Scientific Chemicals
Triethylaluminium, 1.3M solution in Heptane, Thermo Scientific Chemicals
Triethylaluminium, 1.3M solution in Heptane, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylaluminium, 1.3M solution in Heptane, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 377298000
Price (EUR)
-
Quantity:
800 mL
Request bulk or custom format
Chemical Identifiers
CAS97-93-8
IUPAC Nametriethylalumane
Molecular FormulaC6H15Al
InChI KeyVOITXYVAKOUIBA-UHFFFAOYSA-N
SMILESCC[Al](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Strength19.5 to 20.5 wt % (on Al basis)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only