Diethylzinc, 1.5M solution in toluene, AcroSeal™
Diethylzinc, 1.5M solution in toluene, AcroSeal™
Diethylzinc, 1.5M solution in toluene, AcroSeal™
Thermo Scientific Chemicals

Diethylzinc, 1.5M solution in toluene, AcroSeal™

CAS: 557-20-0 | C4H10Zn | 123.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 377311000
Price (USD)
165.65
Online exclusive
184.00
Save 18.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
165.65
Online exclusive
184.00
Save 18.35
Each
Chemical Identifiers
CAS557-20-0
IUPAC Namediethylzinc
Molecular FormulaC4H10Zn
InChI KeyHQWPLXHWEZZGKY-UHFFFAOYSA-N
SMILESCC[Zn]CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration Complexometric1.4 to 1.6 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only