Methylmagnesium bromide, 1M to 1.5M solution in THF, AcroSeal™
Methylmagnesium bromide, 1M to 1.5M solution in THF, AcroSeal™
Methylmagnesium bromide, 1M to 1.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Methylmagnesium bromide, 1M to 1.5M solution in THF, AcroSeal™

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 377381000
Price (USD)
69.70
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
69.70
Each
Chemical Identifiers
CAS75-16-1
IUPAC Namebromo(methyl)magnesium
Molecular FormulaCH3BrMg
InChI KeyAVFUHBJCUUTGCD-UHFFFAOYSA-M
SMILESC[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to turbid grey to dark purple to brown-black
Appearance (Form)Solution
Titration Grignard0.95 to 1.50 M
AppearanceMay develop some turbidity or precipitate
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only