Potassium thiocyanate, 98%, pure
Potassium thiocyanate, 98%, pure
Potassium thiocyanate, 98%, pure
Potassium thiocyanate, 98%, pure
Thermo Scientific Chemicals

Potassium thiocyanate, 98%, pure

CAS: 333-20-0 | CKNS | 97.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 377431000
Price (EUR)
33,10
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
33,10
Each
Add to cart
Chemical Identifiers
CAS333-20-0
IUPAC Namepotassium cyanosulfanide
Molecular FormulaCKNS
InChI KeyZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES[K+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only