(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™
(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™
(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™
Thermo Scientific Chemicals

(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™

CAS: 1822-00-0 | C4H11LiSi | 94.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 377458000
Price (USD)
-
Quantity:
800 mL
Request bulk or custom format
Chemical Identifiers
CAS1822-00-0
IUPAC Namelithium(1+) (trimethylsilyl)methanide
Molecular FormulaC4H11LiSi
InChI KeyKVWLUDFGXDFFON-UHFFFAOYSA-N
SMILES[Li+].C[Si](C)(C)[CH2-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Colorless to yellow-brown
Assay7 to 14 % w/w
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only