(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™
(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™
(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™
Thermo Scientific Chemicals

(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™

CAS: 13170-43-9 | C4H11ClMgSi | 146.97 g/mol
Have Questions?
Catalog NumberQuantityPackaging
377461000100 mLAcroSeal™ Glass bottle
Catalog number 377461000
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS13170-43-9
IUPAC Name[(chloromagnesio)methyl]trimethylsilane
Molecular FormulaC4H11ClMgSi
InChI KeyNAQATMJWCJCHOZ-UHFFFAOYSA-M
SMILESC[Si](C)(C)C[Mg]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear to hazy grey to brown-black
Titration Grignard1.2 to 1.5 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only