tert-Butoxybis(dimethylamino)methane
tert-Butoxybis(dimethylamino)methane
tert-Butoxybis(dimethylamino)methane
Thermo Scientific Chemicals

tert-Butoxybis(dimethylamino)methane

CAS: 5815-08-7 | C9H22N2O | 174.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
37812025025 gGlass bottle
Catalog number 378120250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5815-08-7
IUPAC Name[(tert-butoxy)(dimethylazaniumyl)methyl]dimethylazanium
Molecular FormulaC9H24N2O
InChI KeyHXRAMSFGUAOAJR-UHFFFAOYSA-P
SMILESC[NH+](C)C(OC(C)(C)C)[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC90 % (sum of 3 constituents) varying proportionally
GCTris(dimethylamino)methane
GCtert.-Butoxybis(dimethylamino)methane
Appearance (Color)Clear light yellow
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only