SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Thermo Scientific™
SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks
The pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoVolumen (métrico)Esterilidad
3781RC-HR1000 μLEstéril
3721RC-HR20 μLEstéril
3722RC-HR20 μLNo estéril
3731RC-HR300 μLEstéril
3732RC-HR300 μLNo estéril
3741RC-HR1200 μLEstéril
3742RC-HR1200 μLNo estéril
3751RC-HR200 μLEstéril
3752RC-HR200 μLNo estéril
3782RC-HR1000 μLNo estéril
Número de catálogo 3781RC-HR
Precio (USD)
-
Volumen (métrico):
1000 μL
Esterilidad:
Estéril
Gain the accuracy and precision you expect from your pipette samples every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
Especificaciones
Declaraciones de limpiezaSin ADNasa, ARNasa, ADN, ATP, inhibidor de PCR ni endotoxinas
Incluye1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Longitud (imperial)3,39 in
Longitud (métrico)8.61 cm
Modelo1000 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Tipo de productoTipCycle, Pipette Tip and Rack Recycling Kit
Esterilizables en autoclaveNo Autoclavable
ColorNatural
Código de colorAzul
filtradas porSin filtro
FormatoHinged Rack
Línea de productosSoftfit-L
EsterilidadEstéril
Estilo de puntaSpecialty
Volumen (métrico)1000 μL
Unit SizeCase of 3072