Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™
Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™
Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™
Thermo Scientific Chemicals

Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™

CAS: 7646-78-8 | Cl4Sn | 260.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 378201000
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
72.00
Save 10.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
72.00
Save 10.35
Each
Chemical Identifiers
CAS7646-78-8
IUPAC Nametin(IV) chloride
Molecular FormulaCl4H4Sn
InChI KeyHTGSJGUTBKFNJC-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric0.95 to 1.10 M
Appearance (Color)Clear colorless
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only