SoftFit-L™ Non-Filtered Pipette Tips in Racks with Lift-off Lid

SoftFit-L™ Non-Filtered Pipette Tips in Racks with Lift-off Lid

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
3782TSPK
also known as 3782
1000 μLNon-sterile
3721TSPK
also known as 3721
20 μLSterile
3751TSPK
also known as 3751
200 μLSterile
3752TSPK
also known as 3752
200 μLNon-sterile
3781TSPK
also known as 3781
1000 μLSterile
Catalog number 3782TSPK
also known as 3782
Price (MXN)
-
Volume (Metric):
1000 μL
Sterility:
Non-sterile
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.

SoftFit-L tips have thinner walls than conventional tips so they conform more easily to the pipetter barrel, which ensures a better seal and allows the tip to eject with less force.

  • Non-Filtered tips are available in sterile and non-sterile
  • Available racked or in hinged racks
Hinged Racks
  • Covers can be used as hinged or lift-off style
  • Easy, one-hand operation of locking clasp
  • Pipet tip inserts clip securely into racks but are easy to remove
  • Empty rack part numbers allow for easy conversion to Reload System
  • All components are completely recyclable and contain recycling symbols
  • Economical and environmental design reduces waste and saves bench space
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description1000L, Non-Filtered, Non-sterile, Rack
Length (English)3.39 in.
Length (Metric)8.61cm
Model1000L
ColorNatural
Color CodeBlue
FilteredNon-filtered
For Use With (Equipment)Rainin LTS LiteTouch
FormatRack
Product LineART SoftFit L
SterilityNon-sterile
Tip StyleUniversal
Volume (Metric)1000 μL
Unit SizePack of 768