Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™
Thermo Scientific Chemicals

Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™

CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 378411000
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS74-88-4
IUPAC Nameiodomethane
Molecular FormulaCH3I
InChI KeyINQOMBQAUSQDDS-UHFFFAOYSA-N
SMILESCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
StabilizerSilver
Appearance (Color)Clear colorless to light yellow
Strength1.90 to 2.20 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only