N-(5-chloro-2-pyridyl)bis(trifluoromethanesulfonimide)
N-(5-chloro-2-pyridyl)bis(trifluoromethanesulfonimide)
N-(5-chloro-2-pyridyl)bis(trifluoromethanesulfonimide)
Thermo Scientific Chemicals

N-(5-chloro-2-pyridyl)bis(trifluoromethanesulfonimide)

CAS: 145100-51-2 | C7H3ClF6N2O4S2 | 392.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 378800050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS145100-51-2
IUPAC NameN-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide
Molecular FormulaC7H3ClF6N2O4S2
InChI KeyTUFGVZMNGTYAQD-UHFFFAOYSA-N
SMILESFC(F)(F)S(=O)(=O)N(C1=CC=C(Cl)C=N1)S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline solid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only