Tris(2-methoxyphenyl)phosphine, 97+%
Tris(2-methoxyphenyl)phosphine, 97+%
Tris(2-methoxyphenyl)phosphine, 97+%
Thermo Scientific Chemicals

Tris(2-methoxyphenyl)phosphine, 97+%

CAS: 4731-65-1 | C21H21O3P | 352.37 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 378850010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4731-65-1
IUPAC Nametris(2-methoxyphenyl)phosphane
Molecular FormulaC21H21O3P
InChI KeyIIOSDXGZLBPOHD-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1P(C1=CC=CC=C1OC)C1=CC=CC=C1OC
さらに表示
Infrared spectrumConforms
Melting point192°C to 207°C
HPLC>=97.0 %
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only