Dibutylboron trifluoromethanesulfonate, 1M sol. in dichloromethane
Dibutylboron trifluoromethanesulfonate, 1M sol. in dichloromethane
Dibutylboron trifluoromethanesulfonate, 1M sol. in dichloromethane
Thermo Scientific Chemicals

Dibutylboron trifluoromethanesulfonate, 1M sol. in dichloromethane

CAS: 60669-69-4 | C9H18BF3O3S | 274.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass Bottle
Catalog number 378990250
Price (USD)
98.80
Each
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
98.80
Each
Chemical Identifiers
CAS60669-69-4
IUPAC Namedibutylboranyl trifluoromethanesulfonate
Molecular FormulaC9H18BF3O3S
InChI KeyFAVAVMFXAKZTMV-UHFFFAOYSA-N
SMILESCCCCB(CCCC)OS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange
Appearance (Form)Solution
Strength0.9 to 1.1 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only