alpha-Methylhydrocinnamic acid, 98%
alpha-Methylhydrocinnamic acid, 98%
alpha-Methylhydrocinnamic acid, 98%
Thermo Scientific Chemicals

alpha-Methylhydrocinnamic acid, 98%

CAS: 1009-67-2 | C10H12O2 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 379080050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1009-67-2
IUPAC Name2-methyl-3-phenylpropanoic acid
Molecular FormulaC10H12O2
InChI KeyMCIIDRLDHRQKPH-UHFFFAOYNA-N
SMILESCC(CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals or crystalline mass and/or
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only