2-Bromophenylboronic acid, 97%
2-Bromophenylboronic acid, 97%
2-Bromophenylboronic acid, 97%
Thermo Scientific Chemicals

2-Bromophenylboronic acid, 97%

CAS: 244205-40-1 | C6H6BBrO2 | 200.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 379990050
Price (USD)
86.65
Online exclusive
95.60
Save 8.95
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
86.65
Online exclusive
95.60
Save 8.95
Each
Chemical Identifiers
CAS244205-40-1
IUPAC Name(2-bromophenyl)boronic acid
Molecular FormulaC6H6BBrO2
InChI KeyPLVCYMZAEQRYHJ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Titration with NaOH96.0 to 110.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Powder or crystals and/or chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only