Sodium dihydrogen phosphate monohydrate, 98.0-103.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sodium dihydrogen phosphate monohydrate, 98.0-103.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets U.S.P and B.P. Chemical Specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
Catalog number 3802-07
Price (KRW)
-
Quantity:
12 kg

This product is GMP Manufactured and meets U.S.P and B.P. Chemical Specifications.

Specifications
CAS10049-21-5
Chemical Name or MaterialSodium Dihydrogen Phosphate Monohydrate
ColorWhite
Melting Point100°C
Molecular FormulaNaH2PO4·H2O
PackagingPolyethylene pail, screw top
SynonymSodium dihydrogen orthophosphate 1-hydrate, Sodium dihydrogen orthophosphate monohydrate, Sodium dihydrogen phosphate 1-hydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
GradeMulti-Compendial
Molecular Weight (g/mol)137.99
Percent Purity98.0-103.0%
Quantity12 kg
Unit SizeEach