Sodium diformamide, 98%
Sodium diformamide, 98%
Sodium diformamide, 98%
Thermo Scientific Chemicals

Sodium diformamide, 98%

CAS: 18197-26-7 | C2H2NNaO2 | 95.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 380430250
Price (EUR)
212,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
212,00
Each
Chemical Identifiers
CAS18197-26-7
IUPAC Namesodium diformylazanide
Molecular FormulaC2H2NNaO2
InChI KeyQJXDSDLNUKLDBP-UHFFFAOYSA-M
SMILES[Na+].O=C[N-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with HCl>=97.5 %
Infrared spectrumConforms
Appearance (Color)Light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only