Thermo Scientific Chemicals

N,N-Diformylacetamide, 80%, Thermo Scientific Chemicals

Catalog number: 380450050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Diformylacetamide, 80%, Thermo Scientific Chemicals

Catalog number: 380450050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 380450050
Price (USD)
Price: 249.00
Online price: 215.65
Your price:
Quantity
-

Chemical Identifiers

CAS
26944-31-0
IUPAC Name
N,N-diformylacetamide
Molecular Formula
C4H5NO3
InChI Key
ZAHBLOGBMDVRDK-UHFFFAOYSA-N
SMILES
CC(=O)N(C=O)C=O
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=75 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text