Propargyl acetate, 97%
Propargyl acetate, 97%
Propargyl acetate, 97%
Thermo Scientific Chemicals

Propargyl acetate, 97%

CAS: 627-09-8 | C5H6O2 | 98.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 380820250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS627-09-8
IUPAC Nameprop-2-yn-1-yl acetate
Molecular FormulaC5H6O2
InChI KeyRIZZXCJMFIGMON-UHFFFAOYSA-N
SMILESCC(=O)OCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4165 to 1.4195 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only