4-tert-Butylphenylacetylene, 96%
4-tert-Butylphenylacetylene, 96%
4-tert-Butylphenylacetylene, 96%
Thermo Scientific Chemicals

4-tert-Butylphenylacetylene, 96%

CAS: 772-38-3 | C12H14 | 158.24 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 380850050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS772-38-3
IUPAC Name1-tert-butyl-4-ethynylbenzene
Molecular FormulaC12H14
InChI KeyZSYQVVKVKBVHIL-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to yellow-brown
Infrared spectrumConforms
Refractive index1.5270 to 1.5310 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only