Methyl 5-hexynoate, 95%
Methyl 5-hexynoate, 95%
Methyl 5-hexynoate, 95%
Thermo Scientific Chemicals

Methyl 5-hexynoate, 95%

CAS: 77758-51-1 | C7H10O2 | 126.15 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 380880250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS77758-51-1
IUPAC Namemethyl hex-5-ynoate
Molecular FormulaC7H10O2
InChI KeyLZULAZTXJLWELL-UHFFFAOYSA-N
SMILESCOC(=O)CCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
GC>=94 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only