Lithium Bis(trifluoromethanesulfonimide), 99%
Lithium Bis(trifluoromethanesulfonimide), 99%
Lithium Bis(trifluoromethanesulfonimide), 99%
Thermo Scientific Chemicals

Lithium Bis(trifluoromethanesulfonimide), 99%

CAS: 90076-65-6 | C2F6LiNO4S2 | 287.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 381030100
Price (USD)
93.20
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
93.20
Each
Chemical Identifiers
CAS90076-65-6
IUPAC Namelithium(1+) trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Molecular FormulaC2F6LiNO4S2
InChI KeyQSZMZKBZAYQGRS-UHFFFAOYSA-N
SMILES[Li+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Titration after Ion exchange>=98.5 %
Appearance (Color)Colorless to white
Infrared spectrumConforms
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only