5-Bromo-1-methylimidazole, 97%
5-Bromo-1-methylimidazole, 97%
5-Bromo-1-methylimidazole, 97%
Thermo Scientific Chemicals

5-Bromo-1-methylimidazole, 97%

CAS: 1003-21-0 | C4H5BrN2 | 161.00 g/mol
Have Questions?
Catalog NumberQuantity
3810400505 g
Catalog number 381040050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1003-21-0
IUPAC Name5-bromo-1-methyl-1H-imidazole
Molecular FormulaC4H5BrN2
InChI KeyHATLLUIOEIXWGD-UHFFFAOYSA-N
SMILESCN1C=NC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point38°C to 43°C
GC>=96.0 %
Appearance (Color)White to tan
Appearance (Form)Crystalline solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only