2-(BOC-Amino)ethanethiol, 97%
2-(BOC-Amino)ethanethiol, 97%
2-(BOC-Amino)ethanethiol, 97%
Thermo Scientific Chemicals

2-(BOC-Amino)ethanethiol, 97%

CAS: 67385-09-5 | C7H15NO2S | 177.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
38108025025 mLGlass Bottle
Catalog number 381080250
Price (USD)
242.65
Special offer
Online exclusive
Ends: 28-Dec-2026
285.00
Save 42.35
Each
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
242.65
Special offer
Online exclusive
Ends: 28-Dec-2026
285.00
Save 42.35
Each
Chemical Identifiers
CAS67385-09-5
IUPAC Nametert-butyl N-(2-sulfanylethyl)carbamate
Molecular FormulaC7H15NO2S
InChI KeyGSJJCZSHYJNRPN-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or low melting solid
Refractive index1.4740 to 1.4770 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only