Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™
Thermo Scientific Chemicals

Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™

CAS: 96-10-6 | C4H10AlCl | 120.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 381151000
Price (EUR)
58,65
Online exclusive
65,50
Save 6,85
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
58,65
Online exclusive
65,50
Save 6,85
Each
Chemical Identifiers
CAS96-10-6
IUPAC Namechlorodiethylalumane
Molecular FormulaC4H10AlCl
InChI KeyYNLAOSYQHBDIKW-UHFFFAOYSA-M
SMILESCC[Al](Cl)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Strength11.5 to 12.5 % (on Al basis)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only