Triethylaluminium, 0.6M solution in heptane, AcroSeal™
Triethylaluminium, 0.6M solution in heptane, AcroSeal™
Triethylaluminium, 0.6M solution in heptane, AcroSeal™
Thermo Scientific Chemicals

Triethylaluminium, 0.6M solution in heptane, AcroSeal™

CAS: 97-93-8 | C6H15Al | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 381178000
Price (USD)
-
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS97-93-8
IUPAC Nametriethylalumane
Molecular FormulaC6H15Al
InChI KeyVOITXYVAKOUIBA-UHFFFAOYSA-N
SMILESCC[Al](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Strength9.5 to 10.5 wt % (on Al basis)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only