Appearance (Form)Crystalline powder or flakes and/or chunks
Infrared spectrumConforms
Appearance (Color)White to light yellow
Melting point122°C to 126°C
Titration by coupling>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.