Cyclopentylacetylene, 95%
Cyclopentylacetylene, 95%
Cyclopentylacetylene, 95%
Thermo Scientific Chemicals

Cyclopentylacetylene, 95%

CAS: 930-51-8 | C7H10 | 94.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 381310050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS930-51-8
IUPAC Nameethynylcyclopentane
Molecular FormulaC7H10
InChI KeyTXVJSWLZYQMWPC-UHFFFAOYSA-N
SMILESC#CC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Refractive index1.4470 to 1.4490 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only