2-(Trimethylsilylmethyl)allyl acetate, 94%
2-(Trimethylsilylmethyl)allyl acetate, 94%
2-(Trimethylsilylmethyl)allyl acetate, 94%
Thermo Scientific Chemicals

2-(Trimethylsilylmethyl)allyl acetate, 94%

CAS: 72047-94-0 | C9H18O2Si | 186.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 381380250
Price (EUR)
971,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
971,00
Each
Chemical Identifiers
CAS72047-94-0
IUPAC Name2-[(trimethylsilyl)methyl]prop-2-en-1-yl acetate
Molecular FormulaC9H18O2Si
InChI KeyIKQWABMHZKGCLX-UHFFFAOYSA-N
SMILESCC(=O)OCC(=C)C[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=92 %
Refractive index1.4380 to 1.4420 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only