Disodium hydrogen phosphate heptahydrate, 98.0-100.5% (dried basis), crystals, USP, endotoxin tested, GMP, J.T.Baker™
Avantor™

Disodium hydrogen phosphate heptahydrate, 98.0-100.5% (dried basis), crystals, USP, endotoxin tested, GMP, J.T.Baker™

This product is GMP Manufactured and meets U.S.P Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
12 kg
200 lb.
Catalog number 3816-07
Price (MXN)
-
Quantity:
12 kg
This product is GMP Manufactured and meets U.S.P Requirements.
Specifications
CAS7782-85-6
ColorWhite
Melting Point487°C
Molecular FormulaHna2O4P·7H2O
PackagingPolyethylene pail, screw top
SynonymSodium phosphate dibasic heptahydrate, Sodium monohydrogen phosphate heptahydrate, Disodium hydrogenorthophosphate 7-hydrate, Disodium hydrogenorthophosphate heptahydrate, Sodium phosphate dibasic 7-hydrate
GradeUSP
Molecular Weight (g/mol)268.07
Percent Purity98.0-100.5%
Quantity12 kg
Unit SizeEach