Nalgene™ Teflon™ Low Particulate Bottles
Actual product may vary
Thermo Scientific™

Nalgene™ Teflon™ Low Particulate Bottles

Nalgene Teflon Low Particulate Bottles are certified clean to USP <788> standards, making them an ideal solution for biological and pharmaceutical manufacturing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
381600-0008250 mL24 mm
381600-0004125 mL24 mm
381600-0016500 mL28 mm
381600-00321 L38 mm
Catalog number 381600-0008
Price (BRL)
-
Capacity (Metric):
250 mL
Closure Size:
24 mm

Sample and store active pharmaceutical ingredients and bulk intermediates optimally with Thermo Scientific™ Nalgene™ Low Particulate/Low Metals Bottles Made of Teflon™ FEP with Closure, certified to contain less than 20 particles per mL at 0.3 μm and greater. Metals certified to micrograms per liter (ppb) levels. Excellent for storing high purity chemicals, this product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.

Features:

  • Certified to particulate and trace metals levels
  • Linerless Tefzel™ ETFE closures provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks, or contaminate your reagents
  • Exhibits flexibility, impact resistance and excellent visibility of contents for a wide variety of uses in the lab
  • Excellent chemical resistance of almost any lab chemical
  • Excellent for storing high purity chemicals
  • Non-sterile/autoclavable
  • Translucent

Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. Excellent for ICP-MS reagent and standard storage. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.

Metals certified to microliter (ppb) levels: <0.20 Hg, <0.5 Be, <1.0 As/Cd/Pb, <2.0 Sb/Se, <5.0 Ag/Co/Cr/Cu/Mn/Tl/V, <10 Ba/Ni/Zn, <50 Mg, <75 Al, <100 Ca/Fe/K/Na

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableAutoclavable
Capacity (English)8 oz.
Certifications/ComplianceCertified to particulate and trace metals levels. Metals certified to microliter (ppb) levels <0.20 Hg, <0.5 Be, <1.0 As/Cd/Pb, <2.0 Sb/Se, <5.0 Ag/Co/Cr/Cu/Mn/Tl/V, <10 Ba/Ni/Zn, <75 Al, <100 Ca/Fe/K/Na. Certified to contain <20 particles
Closure ColorWhite
Closure LinerUnlined
Closure MaterialTefzel™ ETFE
Closure Size24 mm
Closure TypeScrew Cap
Diameter (Metric)60 mm
Diameter (Metric) Inner Neck17 mm
For Use With (Application)Storage, Transportation ​
FormatLow Particulate/Low Metals Bottle
Height (English)5.27 in.
Height (Metric)134 mm
Low ParticulateUSP <788> (Low particulate)
Neck Finish24 mm
No. per Case4/Cs.
No. per Pack1 Ea.
Product TypeLow Particulate/Low Metals Bottle
ShapeRound
Temperature Range (Metric)-100°C to 260°C
Through the Closure DispensingNo
Width (English)2.36 in.
Workflow StepUpstream process solution storage. Upstream Validation. Downstream process solution storage. Downstream Validation
Capacity (Metric)250 mL
Closure IncludedYes
ColorClear
MaterialTeflon FEP
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 4

Frequently asked questions (FAQs)

Why do you recommend Teflon-coated bottles to store high-value contents such as, active pharmaceutical ingredients (API's), bulk intermediates, and critical reagents?

Teflon is ideal for storing the highest-value contents because it is resistant to most chemicals, temperatures, and processes, allowing for rigorous cleaning procedures such as those that require boiling nitric acid. Teflon also allows you to store contents down to cryogenic temperatures for products requiring ultra-low temperature (below -100 degrees C) storage.
We offer Nalgene Teflon bottles that provide excellent chemical and temperature resistance in a leak-proof design. They are manufactured using quality resins, with the added value of high-level support, making Nalgene Teflon bottles an ideal choice for your high-value contents.