ácido 2-cloro-5-metilbenzoico, 98 %, Thermo Scientific Chemicals
ácido 2-cloro-5-metilbenzoico, 98 %, Thermo Scientific Chemicals
ácido 2-cloro-5-metilbenzoico, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

ácido 2-cloro-5-metilbenzoico, 98 %, Thermo Scientific Chemicals

CAS: 6342-60-5 | C8H7ClO2 | 170.59 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 381970010
Precio (EUR)
47,80
Each
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
47,80
Each
Identificadores químicos
CAS6342-60-5
IUPAC Name2-chloro-5-methylbenzoic acid
Molecular FormulaC8H7ClO2
InChI KeyLEBWXJZAWTVKFL-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C(=C1)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder
Melting point148°C to 151°C
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Off-white to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only