2-chloro-5-methylbenzoic acid, 98%
2-chloro-5-methylbenzoic acid, 98%
2-chloro-5-methylbenzoic acid, 98%
Thermo Scientific Chemicals

2-chloro-5-methylbenzoic acid, 98%

CAS: 6342-60-5 | C8H7ClO2 | 170.59 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 381970050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6342-60-5
IUPAC Name2-chloro-5-methylbenzoic acid
Molecular FormulaC8H7ClO2
InChI KeyLEBWXJZAWTVKFL-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point148°C to 151°C
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Off-white to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only