Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
Thermo Scientific™

Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure

Accurately fill and pour with these narrow-mouth, LDPE low particulate/low metals bottles with closure.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)마개 크기
382003-0008250 mL24 mm
382003-0004125 mL24 mm
382003-0016500 mL28 mm
382003-00321 L38-430 mm
카탈로그 번호 382003-0008
제품 가격(KRW)
-
용량(미터법):
250 mL
마개 크기:
24 mm

Accurately pour and fill Thermo Scientific™ Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure, which feature a narrow mouth. These low-density polyethylene bottles include certification to contain less than 20 particles per mL at 0.3 microns or greater.

Features:
  • Certified to particulate and trace metals levels
  • Excellent chemical resistance to most acids, bases and alcohols
  • Translucent
  • Leakproof† when used with Nalgene closures
Metals certified to microliter (ppb) levels <0.20Hg, <0.5 Be, <1.0 As, Cd, Pb, <2.0 Sb, Se, <5.0 Ag, Co, Cr, Cu, Mn, Tl, V, <10 Ba, Ni, Zn, <50 Mg, <75 Al, <100 Ca, Fe, K, Na. Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. Excellent for ICP-MS reagent and standard storage. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.



Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

사양
오토클레이브 가능Not Autoclavable
용량8 oz., 250 mL
용량(영국식 단위)8 oz.
인증/적합성Certified to particulate and trace metals levels
마개 색상Natural, translucent
마개 라이너Unlined
마개 재질Polypropylene
마개 크기24 mm
마개 유형Screw Cap
설명Low Particulate/Low Metals Bottle
직경(미터법)61 mm
직경(미터법) 내부 넥18 mm
높이(영국식 단위)5.23 in.
높이(미터법)133 mm
저 입자Yes
넥 마감24 mm
케이스당 수72
팩당 수1
제품 유형Low Particulate/Low Metals Bottle
형상Round
마개 디스펜싱을 통해No
폭(영국식 단위)2.4 in.
용량(미터법)250 mL
Closure IncludedYes
색상Translucent
물질LDPE
마우스Narrow
멸균Non-sterile
Unit SizeCase of 72