Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
Thermo Scientific™

Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure

Accurately fill and pour with these narrow-mouth, LDPE low particulate/low metals bottles with closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
382003-0016500 mL28 mm
382003-0004125 mL24 mm
382003-0008250 mL24 mm
382003-00321 L38-430 mm
Catalog number 382003-0016
Price (MXN)
-
Capacity (Metric):
500 mL
Closure Size:
28 mm

Accurately pour and fill Thermo Scientific™ Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure, which feature a narrow mouth. These low-density polyethylene bottles include certification to contain less than 20 particles per mL at 0.3 microns or greater.

Features:
  • Certified to particulate and trace metals levels
  • Excellent chemical resistance to most acids, bases and alcohols
  • Translucent
  • Leakproof† when used with Nalgene closures
Metals certified to microliter (ppb) levels <0.20Hg, <0.5 Be, <1.0 As, Cd, Pb, <2.0 Sb, Se, <5.0 Ag, Co, Cr, Cu, Mn, Tl, V, <10 Ba, Ni, Zn, <50 Mg, <75 Al, <100 Ca, Fe, K, Na. Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. Excellent for ICP-MS reagent and standard storage. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.



Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableAutoclavable
Capacity16 oz., 500 mL
Closure ColorNatural, translucent
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
DescriptionLow Particulate/Low Metals Bottle
Diameter (Metric)73 mm
Diameter (Metric) Inner Neck21 mm
Height (English)6.71 in.
Height (Metric)170 mm
Low ParticulateYes
Neck Finish28 mm
No. per Case48
No. per Pack1
Through the Closure DispensingNo
Width (English)2.86 in.
Capacity (English)16 oz.
Capacity (Metric)500 mL
Certifications/ComplianceCertified to particulate and trace metals levels
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialLDPE
MouthNarrow
Product TypeLow Particulate/Low Metals Bottle
ShapeRound
SterilityNon-sterile
Unit SizeCase of 48