Nalgene™ PETG Low Particulate Media Bottles
Actual product may vary
Thermo Scientific™

Nalgene™ PETG Low Particulate Media Bottles

Nalgene PETG Low Particulate Media Bottles are certified clean to USP <788> standards, making them an ideal solution for biological and pharmaceutical manufacturing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
382019-0250250 mL
382019-003030 mL
382019-006060 mL
382019-0125125 mL
382019-0500500 mL
382019-20002 L
382019-10001 L
Catalog number 382019-0250
Price (TWD)
-
Capacity (Metric):
250 mL

Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.

Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.

  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.

Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableNot Autoclavable
Capacity (English)8.5 oz.
Certifications/ComplianceUSP Class VI, European Pharmacopeia for Abnormal Toxicity and USP 661 physicochemical tests compliant. Manufactured in an ISO 14644-1 Class 7 cleanroom. Certified sterile (SAL 10-6), non-cytotoxic, non-pyrogenic, and non-hemolytic.
Closure Size38-430 mm
Diameter (English)2.4 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric)61 mm
Diameter (Metric) Inner Neck28 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Height (English)5.75 in.
Height (Metric)146 mm
Low ParticulateUSP <788> (Low particulate)
No. per Case48/Cs.
No. per Pack6/Pk.
Product TypeClean Container
ShapeSquare
Temperature Range (Metric)-70°C to 70°C
Workflow StepUpstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
Capacity (Metric)250 mL
Closure IncludedYes
GraduatedYes
MaterialPETG
MouthNarrow
SterilitySterile
Unit SizeCase of 48