Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
Thermo Scientific™

Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack

Effectively store high-purity chemicals with these HDPE bottles, which are extremely low in trace metal content.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
382099-0250250 mL24 mm
382099-0125125 mL24 mm
382099-0500500 mL28 mm
382099-10001 L38-430 mm
Catalog number 382099-0250
Price (USD)
-
Capacity (Metric):
250 mL
Closure Size:
24 mm

Thermo Scientific™ Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles are extremely low in trace metal content, making them an excellent choice for storing high-purity chemicals. Controlled environment manufacturing minimizes particulate contamination. Narrow-mouth bottles with certified low-particulate levels are used by shippers who design and assemble combination packaging for high-purity chemicals.

Features:
  • Leakproof† when used with Nalgene closures

Includes:

Polypropylene closures

Recommended for:

Storage and shipping of microelectronic or biological reagents

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity8 oz., 250 mL
Capacity (English)8 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27(c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Closure ColorNatural
Closure MaterialPolypropylene
Closure Size24 mm
Closure TypeScrew Closure (Assembled)
DescriptionLow-Particulate Narrow-Mouth Bottles
Diameter (Metric)61 mm
Diameter (Metric) Inner Neck18 mm
Height (English)5.23 in.
Height (Metric)133 mm
Low ParticulateYes
Neck Finish24 mm
No. per Case72
Product TypeNarrow Mouth Bottle
ShapeRound
Through the Closure DispensingNo
Capacity (Metric)250 mL
Closure IncludedYes
ColorNatural
MaterialHDPE
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 72