Sodium dihydrogen phosphate monohydrate, 98.0-103.0% (by anhydrous basis), crystals, USP, FCC endotoxin tested, GMP, J.T.Baker™
Sodium dihydrogen phosphate monohydrate, 98.0-103.0% (by anhydrous basis), crystals, USP, FCC endotoxin tested, GMP, J.T.Baker™
Avantor™
Sodium dihydrogen phosphate monohydrate, 98.0-103.0% (by anhydrous basis), crystals, USP, FCC endotoxin tested, GMP, J.T.Baker™
This product is Food GMP Manufactured and meets F.C.C. and USP Requirements.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
수량:
500 g
12 kg
카탈로그 번호 3821-07
제품 가격(KRW)
-
견적 요청하기
수량:
12 kg
This product is Food GMP Manufactured and meets F.C.C. and USP Requirements.
사양
CAS10049-21-5
화학물질 이름 또는 재질Sodium Dihydrogen Phosphate Monohydrate
색상White
용융점100°C
분자식NaH2PO4·H2O
포장Polyethylene pail, screw top
동의어Sodium Phosphate Monobasic hydrate, Sodium dihydrogen orthophosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate 1-hydrate
등급FCC
분자량(g/mol)137.99
백분율 순도98.0-103.0%
수량12 kg
Unit SizeEach