Cellulose microcristalline, extra-pure, granulométrie moyenne 90 μm, Thermo Scientific Chemicals
Cellulose microcristalline, extra-pure, granulométrie moyenne 90 μm, Thermo Scientific Chemicals
Cellulose microcristalline, extra-pure, granulométrie moyenne 90 μm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cellulose microcristalline, extra-pure, granulométrie moyenne 90 μm, Thermo Scientific Chemicals

CAS: 9004-34-6
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
250 g
1 kg
5 kg
25 kg
Conditionnement:
Tout afficher
Flacon en verre
Référence 382312500
Prix (EUR)
61,65
Offre exceptionnelle en ligne
68,80
Économisez 7,15 (10%)
Each
Quantité:
250 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
61,65
Offre exceptionnelle en ligne
68,80
Économisez 7,15 (10%)
Each
Identifiants chimiques
CAS9004-34-6
IUPAC Name2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
Molecular FormulaC14H26O11
InChI KeyPTHCMJGKKRQCBF-UHFFFAOYNA-N
SMILESCOC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O
Voir plus
Appearance (Form)Crystalline or granular powder
Particle size>=70 % (> 32 µm)
Heavy metals=<10 ppm
Appearance (Color)White to off-white
Infrared spectrumConforms
Voir plus
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adjuvant de filtration
RUO – Research Use Only