Sodium phosphate dibasic, 98.0-100.5% (dried basis), USP, GMP, J.T.Baker™
Sodium phosphate dibasic, 98.0-100.5% (dried basis), USP, GMP, J.T.Baker™
Avantor™

Sodium phosphate dibasic, 98.0-100.5% (dried basis), USP, GMP, J.T.Baker™

This product is GMP Manufactured and meets U.S.P Requirements.深入閱讀
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 g
1 kg
12 kg
產品號碼 3827-19
價格 (TWD)
-
申請報價
Quantity:
1 kg
This product is GMP Manufactured and meets U.S.P Requirements.
規格
CAS7558-79-4
Chemical Name or MaterialSodium phosphate dibasic
ColorWhite
Melting Point250°C
Molecular FormulaNa2HPO4
PackagingAmber glass bottle, wide mouth, tamper-evident cap
SynonymSodium phosphate dibasic, Disodium hydrogen orthophosphate, Sodium hydrogen phosphate
GradeUSP
Molecular Weight (g/mol)141.96
Percent Purity98.0-100.5%
Quantity1 kg
Unit SizeEach