Nalgene™ Certified Clean PETG Biotainer™ Media Bottle
Thermo Scientific™
Nalgene™ Certified Clean PETG Biotainer™ Media Bottle
For Use With: Specifically designed for pharmaceutical and biological manufacturing.; Color: Clear
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
383005-70500 mL
383110-351 L
Catalog number 383005-70
Price (BRL)
-
Capacity (Metric):
500 mL
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Media Bottles reduce the risk of carry-over contamination.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AutoclavableNot Autoclavable
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Low ParticulateUSP <788> (Low particulate)
No. per Case70/Cs.
Workflow StepUpstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
Capacity (Metric)500 mL
ColorClear
MaterialPET
SterilitySterile
Unit SizeEach