Sodium hydroxide, for analysis, micropearls
Sodium hydroxide, for analysis, micropearls
Sodium hydroxide, for analysis, micropearls
Thermo Scientific Chemicals

Sodium hydroxide, for analysis, micropearls

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
產品號碼 383040010
價格 (TWD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
Titration with HCl>=99.0 %
Sodium chloride (NaCl)=<200 ppm
Lead (Pb)=<0.5 ppm
Sodium sulfate (Na2SO4)=<80 ppm
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only