Sodium hydroxide, for analysis, micropearls
Sodium hydroxide, for analysis, micropearls
Sodium hydroxide, for analysis, micropearls
Thermo Scientific Chemicals

Sodium hydroxide, for analysis, micropearls

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 383042500
Price (JPY)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HCl>=99.0 %
Sodium chloride (NaCl)=<200 ppm
Lead (Pb)=<0.5 ppm
Sodium sulfate (Na2SO4)=<80 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only